missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPTM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32114
This item is not returnable.
View return policy
Description
CLPTM1 Polyclonal specifically detects CLPTM1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| CLPTM1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| cleft lip and palate associated transmembrane protein 1, cleft lip and palate transmembrane protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1209 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate | |
| NP_001285.1 | |
| CLPTM1 | |
| The immunogen this antibody was made to, maps to a region between residue 619 to 669 of human Cleft Lip and Palate Associated Transmembrane Protein 1 using the numbering given in entry NP_001285.1 (GeneID 1209). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction