missing translation for 'onlineSavingsMsg'
Learn More

CLPTM1 Antibody, Biotin, Novus Biologicals™

Product Code. 18432732 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432732 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432732 Supplier Novus Biologicals Supplier No. NBP232114B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CLPTM1 Polyclonal antibody specifically detects CLPTM1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPTM1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias cleft lip and palate associated transmembrane protein 1, cleft lip and palate transmembrane protein 1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 619 to 669 of human Cleft Lip and Palate Associated Transmembrane Protein 1 using the numbering given in entry NP_001285.1 (GeneID 1209).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 1209
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.