missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPS Antibody (111), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90233R
This item is not returnable.
View return policy
Description
CLPS Monoclonal specifically detects CLPS in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CLPS | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 111 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| colipase, colipase, pancreatic, pancreatic colipase preproprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CLPS (P04118; Met1-Gln112). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1208 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction