missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLPP Recombinant Rabbit Monoclonal Antibody (1W6O4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538052
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, SW620, HepG2, Mouse liver, Mouse heart, Rat liver Immunogen sequence: HQPSGGARGQ ATDIAIQAEE IMKLKKQLYN IYAKHTKQSL QVIESAMERD RYMSPMEAQE FGILDKVLVH PPQDGEDEPT LVQKEPVEAA PAAEPVPAST.
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
Specifications
| CLPP | |
| Recombinant Monoclonal | |
| 0.46 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88696, Q16740 | |
| CLPP | |
| A synthetic peptide corresponding to a sequence within amino acids 178-277 of human CLPP (Q16740). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1W6O4 | |
| Unconjugated | |
| CLPP | |
| ATP-dependent Clp protease proteolytic subunit, mitochondrial; ATP-dependent protease ClpAP, proteolytic subunit, human; AU019820; caseinolytic mitochondrial matrix peptidase proteolytic subunit; caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; caseinolytic protease, ATP-dependent, proteolytic subunit homolog; Clpp; ClpP caseinolytic peptidase ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog; D17Wsu160e; DFNB81; endopeptidase Clp; LOW QUALITY PROTEIN: ATP-dependent Clp protease proteolytic subunit, mitochondrial; PRLTS3; putative ATP-dependent Clp protease proteolytic subunit, mitochondrial | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 301117, 53895, 8192 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction