missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15698-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.Specifications
| CLPP | |
| Polyclonal | |
| Unconjugated | |
| CLPP | |
| CLPP, Endopeptidase Clp | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 301117, 53895, 8192 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88696, Q16740 | |
| Clpp | |
| CLPP Fusion Protein Ag8325 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction