missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLPP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117875
This item is not returnable.
View return policy
Description
CLPP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
Specifications
| CLPP | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q16740 | |
| CLPP | |
| E. coli-derived Human CLPP fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| CLPP | |
| ATP-dependent Clp protease proteolytic subunit, mitochondrial; ATP-dependent protease ClpAP, proteolytic subunit, human; AU019820; caseinolytic mitochondrial matrix peptidase proteolytic subunit; caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; caseinolytic protease, ATP-dependent, proteolytic subunit homolog; Clpp; ClpP caseinolytic peptidase ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit; ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog; ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog; D17Wsu160e; DFNB81; endopeptidase Clp; LOW QUALITY PROTEIN: ATP-dependent Clp protease proteolytic subunit, mitochondrial; PRLTS3; putative ATP-dependent Clp protease proteolytic subunit, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 8192 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction