missing translation for 'onlineSavingsMsg'
Learn More

CLPP Antibody, DyLight 650, Novus Biologicals™

Product Code. 18381305 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381305 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381305 Supplier Novus Biologicals Supplier No. NBP306239C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CLPP Polyclonal specifically detects CLPP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLPP
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent protease ClpAP, proteolytic subunit, human, ClpP (caseinolytic protease, ATP-dependent, proteolytic subunit, E. coli)homolog, ClpP caseinolytic peptidase, ATP-dependent, proteolytic subunit homolog (E.coli), ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog, ClpP caseinolytic protease, ATP-dependent, proteolytic subunit homolog (E.coli), EC 3.4.21.92, Endopeptidase Clp, putative ATP-dependent Clp protease proteolytic subunit, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CLPP fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8192
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.