missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CLPB |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16875054
|
Proteintech
15743-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16865054
|
Proteintech
15743-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLPB may function as a regulatory ATPase and be related to secretion/protein trafficking process.Specifications
| CLPB | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q60649, Q9H078, Q9WTT2 | |
| 20480, 65041, 81570 | |
| CLPB Fusion Protein Ag8165 | |
| Primary | |
| -20°C | |
| CLPB |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CLPB, HSP78, SKD3 | |
| Clpb | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title