missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: RRKENGWYDE EHPLVFLFLG SSGIGKTELA KQTAKYMHKD AKKGFIRLDM SEFQERHEVA KFIGSPPGYV GHEEGGQLTK KLKQCPNAVV LF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
CLPB may function as a regulatory ATPase and be related to secretion/protein trafficking process.
Specifications
Specifications
| Antigen | CLPB |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Clpb |
| Gene Accession No. | Q9H078 |
| Gene Alias | AL118244; ANKCLB; ankyrin-repeat containing bacterial clp fusion; caseinolytic mitochondrial matrix peptidase chaperone subunit B; caseinolytic peptidase B; caseinolytic peptidase B protein homolog; CLPB; ClpB caseinolytic peptidase B; ClpB caseinolytic peptidase B homolog; ClpB caseinolytic peptidase B homolog (E. coli); ClpB homolog, mitochondrial AAA ATPase chaperonin; HSP78; MEGCANN; MGCA7; Skd3; suppressor of K+ transport defect 3; suppressor of potassium transport defect 3; testicular secretory protein Li 11 |
| Gene Symbols | Clpb |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
