missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57702
This item is not returnable.
View return policy
Description
CLPB Polyclonal specifically detects CLPB in Human samples. It is validated for Western Blot.
Specifications
| CLPB | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| caseinolytic peptidase B protein homolog, ClpB caseinolytic peptidase B homolog (E. coli), HSP78SKD3FLJ13152, Suppressor of potassium transport defect 3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Canine: 92%; Equine: 92%; Mouse: 91%; Rat: 91%; Bovine: 85%; Rabbit: 84%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9H078 | |
| CLPB | |
| Synthetic peptides corresponding to CLPB(ClpB caseinolytic peptidase B homolog (E. coli)) The peptide sequence was selected from the N terminal of CLPB. Peptide sequence QGGRFDTKCLAAATWGRLPGPEETLPGQDSWNGVPSRAGLGMCALAAALV. | |
| 100 μL | |
| Protein Phosphatase | |
| 81570 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction