missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPB Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97248AF405
This item is not returnable.
View return policy
Description
CLPB Polyclonal antibody specifically detects CLPB in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CLPB | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| caseinolytic peptidase B protein homolog, ClpB caseinolytic peptidase B homolog (E. coli), HSP78SKD3FLJ13152, Suppressor of potassium transport defect 3 | |
| Produced in rabbits immunized with E. coli-derived Human CLPB fragment. | |
| 0.1 mL | |
| Protein Phosphatase | |
| 81570 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction