missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14746-1-AP-20UL
This item is not returnable.
View return policy
Description
HEAB is a protein coding gene.Specifications
| CLP1 | |
| Polyclonal | |
| Unconjugated | |
| CLP1 | |
| CLP1, hClp1, HEAB, Polynucleotide kinase Clp1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10978, 98985 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92989, Q99LI9 | |
| Clp1 | |
| CLP1 Fusion Protein Ag6479 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction