missing translation for 'onlineSavingsMsg'
Learn More

CLP SVOC Calibration Mixture 512 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30328290
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328290 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328290 Supplier Dr. Ehrenstorfer Supplier No. DREA50000512DI
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylnaphthalene∣2-Methylphenol∣2-Nitroaniline∣2-Nitrophenol∣3-Nitroaniline∣4-Chloro-3-methylphenol∣4-Chloroaniline∣4-Chlorophenyl phenyl ether∣4-Methylphenol (p-Cresol)∣4-Nitroaniline∣4-Nitrophenol∣Acenaphthene∣Acenaphthylene∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Carbazole∣Chrysene∣Dibenzo(a,h)anthracene∣Dibenzofuran∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Pentachlorophenol∣Phenanthrene∣Phenol∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene
CAS 120-82-1, 95-50-1, 541-73-1, 106-46-7, 95-95-4, 88-06-2
Analyte or Component Names 1,2,4-Trichlorobenzene∣1,2-Dichlorobenzene∣1,3-Dichlorobenzene∣1,4-Dichlorobenzene∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,4-Dinitrotoluene∣2,6-Dinitrotoluene∣2-Chloronaphthalene∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylnaphthalene∣2-Methylphenol∣2-Nitroaniline∣2-Nitrophenol∣3-Nitroaniline∣4-Chloro-3-methylphenol∣4-Chloroaniline∣4-Chlorophenyl phenyl ether∣4-Methylphenol (p-Cresol)∣4-Nitroaniline∣4-Nitrophenol∣Acenaphthene∣Acenaphthylene∣Anthracene∣Azobenzene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Bis-(2-chloro-1-methylethyl)ether∣Bis(2-chloroethoxy)methane∣Bis-(2-chloroethyl)ether∣Carbazole∣Chrysene∣Dibenzo(a,h)anthracene∣Dibenzofuran∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Fluoranthene∣Fluorene∣Hexachlorobenzene∣Hexachlorobutadiene∣Hexachlorocyclopentadiene∣Hexachloroethane∣Indeno[1,2,3-cd]pyrene∣Isophorone∣Naphthalene∣Nitrobenzene∣N-Nitrosodimethylamine∣N-Nitroso-di-n-propylamine∣PBDE 3 (4-Bromodiphenyl Ether)∣Pentachlorophenol∣Phenanthrene∣Phenol∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester∣Pyrene
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,2,4-Trichlorobenzene 2000 μg/mL∣1,2-Dichlorobenzene 2000 μg/mL∣1,3-Dichlorobenzene 2000 μg/mL∣1,4-Dichlorobenzene 2000 μg/mL∣2,4,5-Trichlorophenol 2000 μg/mL∣2,4,6-Trichlorophenol 2000 μg/mL∣2,4-Dichlorophenol 2000 μg/mL∣2,4-Dimethylphenol 2000 μg/mL∣2,4-Dinitrophenol 2000 μg/mL∣2,4-Dinitrotoluene 2000 μg/mL∣2,6-Dinitrotoluene 2000 μg/mL∣2-Chloronaphthalene 2000 μg/mL∣2-Chlorophenol 2000 μg/mL∣2-Methyl-4,6-dinitrophenol 2000 μg/mL∣2-Methylnaphthalene 2000 μg/mL∣2-Methylphenol 2000 μg/mL∣2-Nitroaniline 2000 μg/mL∣2-Nitrophenol 2000 μg/mL∣3-Nitroaniline 2000 μg/mL∣4-Chloro-3-methylphenol 2000 μg/mL∣4-Chloroaniline 2000 μg/mL∣4-Chlorophenyl phenyl ether 2000 μg/mL∣4-Methylphenol (p-Cresol) 2000 μg/mL∣4-Nitroaniline 2000 μg/mL∣4-Nitrophenol 2000 μg/mL∣Acenaphthene 2000 μg/mL∣Acenaphthylene 2000 μg/mL∣Anthracene 2000 μg/mL∣Azobenzene 2000 μg/mL∣Benz[a]anthracene 2000 μg/mL∣Benzo(k)fluoranthene 2000 μg/mL∣Benzo[a]pyrene 2000 μg/mL∣Benzo[b]fluoranthene 2000 μg/mL∣Benzo[ghi]perylene 2000 μg/mL∣Bis-(2-chloro-1-methylethyl)ether 2000 μg/mL∣Bis(2-chloroethoxy)methane 2000 μg/mL∣Bis-(2-chloroethyl)ether 2000 μg/mL∣Carbazole 2000 μg/mL∣Chrysene 2000 μg/mL∣Dibenzo(a,h)anthracene 2000 μg/mL∣Dibenzofuran 2000 μg/mL∣Dibutyl phthalate 2000 μg/mL∣Diethyl phthalate 2000 μg/mL∣Di-n-octyl phthalate 2000 μg/mL∣Fluoranthene 2000 μg/mL∣Fluorene 2000 μg/mL∣Hexachlorobenzene 2000 μg/mL∣Hexachlorobutadiene 2000 μg/mL∣Hexachlorocyclopentadiene 2000 μg/mL∣Hexachloroethane 2000 μg/mL∣Indeno[1,2,3-cd]pyrene 2000 μg/mL∣Isophorone 2000 μg/mL∣Naphthalene 2000 μg/mL∣Nitrobenzene 2000 μg/mL∣N-Nitrosodimethylamine 2000 μg/mL∣N-Nitroso-di-n-propylamine 2000 μg/mL∣PBDE 3 (4-Bromodiphenyl Ether) 2000 μg/mL∣Pentachlorophenol 2000 μg/mL∣Phenanthrene 2000 μg/mL∣Phenol 2000 μg/mL∣Phthalic acid, benzylbutyl ester 2000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 2000 μg/mL∣Phthalic acid, bis-methyl ester 2000 μg/mL∣Pyrene 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.