missing translation for 'onlineSavingsMsg'
Learn More

Volatiles Mix A, SPEX CertiPrep™

Product Code. 16329887 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 16329887

Brand: SPEX Certiprep CLPVA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,1,2,2-Tetrachloroethane79-34-5200μg/mL
1,1,2-Trichloroethane79-00-5200μg/mL
1,1-Dichloroethane75-34-3200μg/mL
1,1-Dichloroethene75-35-4200μg/mL
1,2-Dichloroethane107-06-2200μg/mL
1,2-Dichloropropane78-87-5200μg/mL
Bromoform75-25-2200μg/mL
Carbon tetrachloride56-23-5200μg/mL
Chlorobenzene108-90-7200μg/mL
Chloroform67-66-3200μg/mL
Dibromochloromethane124-48-1200μg/mL
Methylene chloride75-09-2200μg/mL
Tetrachloroethene127-18-4200μg/mL
trans-1,2-Dichloroethene156-60-5200μg/mL
Trichloroethene79-01-6200μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Volatile Analyte Mix A
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Methods For US EPA Methods 502.1, 502.2, 524, 624, 8260, CLP
Analyte or Component Names Bromoform; Carbon tetrachloride; Chlorobenzene; Chloroform; Dibromochloromethane; 1,1-Dichloroethane; 1,2-Dichloroethane; 1
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Element Bromoform, Carbon Tetrachloride, Chlorobenzene, Chloroform, Dibromochloromethane, 1,1-Dichloroethane, 1,2-Dichloroethane
Solvent or Matrix Methanol-P and T
Quantity 1 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Volatile Analyte Mix A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.