missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLP Instrument Calibration Standard 2: Al, As, Cr, Sb, Se, V @ 1000; Ca, K, Mg, Na @ 10000 μg/mL in 20% HCl, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
CLP Instrument Calibration Standard 2: Al, As, Cr, Sb, Se, V @ 1000; Ca, K, Mg, Na @ 10000 μg/mL in 20% HCl
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Arsenic/Chromium/Selenium/Vanadium/Calcium/Magnesium/Potassium/Sodium |
| CAS | 7429-90-5, 7440-36-0, 7440-38-2, 7440-47-3, 7782-49-2, 7440-62-2 |
| Methods | ICP |
| Analyte or Component Names | Aluminium/Antimony/Arsenic/Chromium/Selenium/Vanadium/Calcium/Magnesium/Potassium/Sodium |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HCl |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 1000 μg/mL/Antimony 1000 μg/mL/Arsenic 1000 μg/mL/Chromium 1000 μg/mL/Selenium 1000 μg/mL/Vanadium 1000 μg/mL/Calcium 10000 μg/mL/Magnesium 10000 μg/mL/Potassium 10000 μg/mL/Sodium 10000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction