missing translation for 'onlineSavingsMsg'
Learn More

CLP Acids Mix 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30309905
Change view
Click to view available options
Quantity:
5 x 5 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309905 5 x 5 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309905 Supplier Dr. Ehrenstorfer Supplier No. DRESY09000023DI
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3,4,6-Tetrachlorophenol∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,6-Dichlorophenol∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylphenol∣2-Nitrophenol∣4-Chloro-3-methylphenol∣4-Methylphenol (p-Cresol)∣4-Nitrophenol∣Benzoic acid∣Pentachlorophenol∣Phenol
CAS 58-90-2, 95-95-4, 88-06-2, 120-83-2, 105-67-9, 51-28-5
Analyte or Component Names 2,3,4,6-Tetrachlorophenol∣2,4,5-Trichlorophenol∣2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2,6-Dichlorophenol∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylphenol∣2-Nitrophenol∣4-Chloro-3-methylphenol∣4-Methylphenol (p-Cresol)∣4-Nitrophenol∣Benzoic acid∣Pentachlorophenol∣Phenol
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Industry Type Food Testing
Quantity 5 x 5 mL
Concentration or Composition (by Analyte or Components) 2,3,4,6-Tetrachlorophenol 1000 μg/mL∣2,4,5-Trichlorophenol 1000 μg/mL∣2,4,6-Trichlorophenol 1000 μg/mL∣2,4-Dichlorophenol 1000 μg/mL∣2,4-Dimethylphenol 1000 μg/mL∣2,4-Dinitrophenol 1000 μg/mL∣2,6-Dichlorophenol 1000 μg/mL∣2-Chlorophenol 1000 μg/mL∣2-Methyl-4,6-dinitrophenol 1000 μg/mL∣2-Methylphenol 1000 μg/mL∣2-Nitrophenol 1000 μg/mL∣4-Chloro-3-methylphenol 1000 μg/mL∣4-Methylphenol (p-Cresol) 1000 μg/mL∣4-Nitrophenol 1000 μg/mL∣Benzoic acid 1000 μg/mL∣Pentachlorophenol 1000 μg/mL∣Phenol 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.