missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CLP 290
Description
CLP 290 is a K+/Cl- Cotransporter 2 (KCC2) activator. Activates spinal inhibitory interneurons, restores physiological neuronal activity in the spinal cord and induces functional recovery in a mouse model of spinal cord injury. Also decreases blood vasopressin and glucose levels in mice with Streptozotocin (Cat. No. 1621) induced diabetes mellitus. Orally bioavailable prodrug of CLP 257 (Cat. No.5242).
Specifications
Specifications
| Chemical Name or Material | (Z)-5-Fluoro-2-((4-oxo-2-(tetrahydropyridazin-1(2 H)-yl)thiazol-5(4 H)-ylidene)methyl)phenyl pyrrolidine-1-carboxylate |
| CAS | 1181083-81-7 |
| Quantity | 50 mg |
| Target | KCC2 Activators |
| Molecular Formula | C19H21FN4O3S |
| Formula Weight | 7828 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?