missing translation for 'onlineSavingsMsg'
Learn More

ClozapineClozapine N-oxide, MedChemExpress

Product Code. 16493317
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493317 50 mg 50mg
16457308 10 mg 10mg
16419817 5 mg 5mg
16467256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493317 Supplier Medchem Express Supplier No. HY17366
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Clozapine N-oxide is a major metabolite of Clozapine and a human muscarinic designer receptors (DREADDs) agonist. Clozapine N-oxide activates the DREADD receptor hM3Dq and hM4Di. Clozapine N-oxide can cross the blood-brain barrier. Clozapine is a potent dopamine antagonist and also a potent and selective muscarinic M4 receptor (EC50=11 nM) agonist.

Clozapine N-oxide is a major metabolite of Clozapine and a human muscarinic designer receptors (DREADDs) agonist. Clozapine N-oxide activates the DREADD receptor hM3Dq and hM4Di. Clozapine N-oxide can cross the blood-brain barrier. Clozapine is a potent dopamine antagonist and also a potent and selective muscarinic M4 receptor (EC50=11 nM) agonist.

Specifications

Chemical Name or Material ClozapineClozapine N-oxide
Physical Form Solid
CAS 34233-69-7
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H19ClN4O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 100 mg/mL (291.70 mM) ∣H2O : 1 mg/mL (2.92 mM; Need ultrasonic)
SMILES [O-][N+]1(C)CCN(C2=NC3=CC(Cl)=CC=C3NC4=CC=CC=C42)CC1
Molecular Weight (g/mol) 342.82
Formula Weight 342.82
Percent Purity 99.25%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.