missing translation for 'onlineSavingsMsg'
Learn More

Clozapine, TRC

Product Code. 18052710
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18052710 250 mg 250mg
30041394 50 mg 50mg
18081500 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18052710 Supplier Toronto Research Chemicals Supplier No. TRCC587500250MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5786-21-0
Molecular Formula C18 H19 Cl N4
Molecular Weight (g/mol) 326.82
Synonym 5H-Dibenzo[b,e][1,4]diazepine, 8-chloro-11-(4-methyl-1-piperazinyl)-, 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]diazepine, Apo-Clozapine, Asaleptin, Azaleptin, Azaleptine, Cloril, Clozapex, Clozapin, Clozapine, Clozaril, Fazaclo, HF 1854, Iprox, Klozapol, Leponex, Sizopin
IUPAC Name 3-chloro-6-(4-methylpiperazin-1-yl)-11H-benzo[b][1,4]benzodiazepine
SMILES CN1CCN(CC1)C2=Nc3cc(Cl)ccc3Nc4ccccc24

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C18H19ClN4/c1-22-8-10-23(11-9-22)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3
Formula Weight 326.1298
Chemical Name or Material Clozapine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.