missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Clozapine N-oxide dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Clozapine N-oxide dihydrochloride is a water-soluble salt of Clozapine N-oxide (Cat. No.4936). Activator of muscarinic DREADDs. Clozapine N-oxide dihydrochloride exhibits improved bioavailability (6 - 7-fold higher plasma concentration) compared with CNO, with less conversion to clozapine in animal studies.
Specifications
Specifications
| Chemical Name or Material | 8-Chloro-11-(4-methyl-4-oxido-1-piperazinyl)-5 H-dibenzo[b,e][1,4 ]diazepine dihydrochloride |
| CAS | 2250025-93-3 |
| Assay Percent Range | >99% |
| Quantity | 50 mg |
| Target | DREADD Activators |
| Molecular Formula | C18H19ClN4O0.2 HCl |
| Formula Weight | Observed MW: 415.74 |
| Purity | 99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction