missing translation for 'onlineSavingsMsg'
Learn More

Cloxyfonac, MedChemExpress

Product Code. 16473407
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473407 50 mg 50mg
16467378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16473407 Supplier Medchem Express Supplier No. HY17527
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cloxyfonac is a plant growth regulator and a chemical transformation product; Pesticide agent.

Cloxyfonac is a plant growth regulator and a chemical transformation product

Specifications

Chemical Name or Material Cloxyfonac
CAS 6386-63-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C9H9ClO4
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (577.05 mM; Need ultrasonic)
SMILES O=C(O)COC1=CC=C(Cl)C=C1CO
Molecular Weight (g/mol) 216.62
Formula Weight 216.62
Percent Purity 99.43%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.