missing translation for 'onlineSavingsMsg'
Learn More

Cloxiquine, MedChemExpress

Product Code. 16411088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
missing translation for 'unitSize':
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
16411088 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16411088 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HYB0963
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cloxiquine (5-Chloro-8-quinolinol) is an antibacterial, antifungal and antiamoebic agent. Cloxiquine can be used for the research of tuberculosis and dermatoses. Cloxiquine suppresses the growth and metastasis of melanoma cells through activation of PPARγ.

Cloxiquine (5-Chloro-8-quinolinol) is an antibacterial, antifungal and antiamoebic agent. Cloxiquine can be used for the research of tuberculosis and dermatoses. Cloxiquine suppresses the growth and metastasis of melanoma cells through activation of PPARγ.

Specifications

Chemical Name or Material Cloxiquine
Physical Form Solid
CAS 130-16-5
Color Light Yellow
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C9H6ClNO
For Use With (Application) COVID-19-immunoregulation
Synonym 5-Chloro-8-quinolinol
Solubility Information DMSO : 100 mg/mL (556.79 mM; Need ultrasonic)
SMILES OC1=C2N=CC=CC2=C(Cl)C=C1
Molecular Weight (g/mol) 179.6
Formula Weight 179.6
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.