missing translation for 'onlineSavingsMsg'
Learn More

Cloxacillin sodium monohydrate

Product Code. 15417889
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417889 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15417889 Supplier Thermo Scientific Alfa Aesar Supplier No. J66500.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7081-44-9 | C19H19ClN3NaO6S | 475.876 g/mol

Targets primarily Gram-positive bacteria, especially --lactamase producing Staphylococci

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Targets primarily gram positive bacteria, especially _x001A_-lactamase producing Staphylococci
TRUSTED_SUSTAINABILITY

Specifications

Type Cloxacillin Sodium Monohydrate
Physical Form Powder
CAS 7081-44-9
Melting Point 170°C
Color White
Quantity 1 g
Flash Point >110°C (230°F)
Molecular Formula C19H19ClN3NaO6S
MDL Number MFCD00150735
Boiling Point 689°C
Merck Index 14,2419
Solubility Information Soluble in water
InChI Key KCUWTKOTPIUBRI-VICXVTCVSA-M
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+]
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
Molecular Weight (g/mol) 475.876
PubChem CID 23675743
ChEBI CHEBI:34978
Formula Weight 475.88
Percent Purity 95%
Chemical Name or Material Cloxacillin sodium monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.