missing translation for 'onlineSavingsMsg'
Learn More

Cloxacillin sodium monohydrate, MedChemExpress

Product Code. 16468326
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468326 100 mg 100mg
16462146 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468326 Supplier Medchem Express Supplier No. HYB0466
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cloxacillin sodium monohydrate is an orally active antibacterial agent and β-lactamase inhibitor with an IC50 of 0.04 μM. Cloxacillin sodium monohydrate can suppress the S. aureus-induced inflammatory response by inhibiting the activation of MAPKs, NF-κB and NLRP3-related proteins.

Cloxacillin sodium monohydrate is an orally active antibacterial agent and β-lactamase inhibitor with an IC50 of 0.04 μM. Cloxacillin sodium monohydrate can suppress the S. aureus-induced inflammatory response by inhibiting the activation of MAPKs, NF-κB and NLRP3-related proteins.

Specifications

Chemical Name or Material Cloxacillin sodium monohydrate
Physical Form Solid
CAS 7081-44-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H19ClN3NaO6S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (210.14 mM; Need ultrasonic) ∣H2O : 50 mg/mL (105.07 mM; Need ultrasonic)
SMILES O=C(C(C(C(C=CC=C1)=C1Cl)=NO2)=C2C)N[C@H]3[C@](SC(C)(C)[C@@H]4C(O[Na])=O)([H])N4C3=O.O
Molecular Weight (g/mol) 475.88
Formula Weight 475.88
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.