missing translation for 'onlineSavingsMsg'
Learn More

Clotrimozole, 99.95%, MP Biomedicals™

Product Code. 11426911 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426911 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11426911 Supplier MP Biomedicals Supplier No. 0219894480
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 23593-75-1
Color White
Assay Percent Range 0.9995
Molecular Formula C22H17ClN2
Quantity 100 g
Synonym clotrimazole, lotrimin, canesten, mycelex, mycosporin, empecid, clotrimazol, mykosporin, gyne lotrimin, chlotrimazole
InChI Key VNFPBHJOKIVQEB-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)N4C=CN=C4
IUPAC Name 1-[(2-chlorophenyl)-diphenylmethyl]imidazole
Molecular Weight (g/mol) 344.842
PubChem CID 2812
ChEBI CHEBI:3764
Formula Weight 344.8
Physical Form Crystalline Powder
Chemical Name or Material Clotrimozole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.