missing translation for 'onlineSavingsMsg'
Learn More

Clotrimazole, LoGiCal

Product Code. 30459919
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30459919 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30459919

Brand: LoGiCal LGCFOR0015.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clotrimazole
CAS 23593-75-1
Analyte or Component Names Clotrimazole
Shipping Condition Room Temperature
Molecular Formula C22 H17 Cl N2
Certifications/Compliance ISO 9001
Synonym Clotrimazole, 1-(o-Chlorophenyldiphenylmethyl)imidazole, 1-(o-Chlorotrityl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole, BAY 5097, BAY 5907, BAY-B 5097, Canesten, Canifug, Desamix F, Diphenyl(2-chlorophenyl)(1-imidazolyl)methane, Empecid, Femcare, Gyne-Lotrimin, Gyne-Lotrimin 7, Lotrimin, Lotrimin AF Cream, Lotrimin AF Solution, Lotrimin Jock-Itch Cream, Lotrimin Jock-Itch Lotion, Monobaycuten, Mycelex, Mycelex 7, Mycelex G, Mycelex OTC, Mycelex Troche, Mycofug, Mycosporin, NSC 257473, Pedisafe, Plimycol, Rimazole, Tibatin, Trimysten, Veltrim, Imidazole, 1-(o-chloro-alpha,alpha-diphenylbenzyl)- (8CI)
InChI Formula InChI=1S/C22H17ClN2/c23-21-14-8-7-13-20(21)22(25-16-15-24-17-25,18-9-3-1-4-10-18)19-11-5-2-6-12-19/h1-17H
Solution Type Neat
SMILES Clc1ccccc1C(c2ccccc2)(c3ccccc3)n4ccnc4
IUPAC Name 1-[(2-chlorophenyl)-diphenylmethyl]imidazole
Industry Type Clinical Testing
Molecular Weight (g/mol) 344.84
Quantity 10 mg
Formula Weight 344.108 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.