missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Closure Ties, Irradiated

Product Code. 18813321 Shop All Micronova Products
Change view
Click to view available options
Dimensions (L x W):
30.48 x 0.58 cm (11.75 x 0.23 in.)
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
18813321 30.48 x 0.58 cm (11.75 x 0.23 in.) Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18813321 Supplier Micronova™ Supplier No. BLT12IR
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Irradiated closure ties are a convenient way to seal liners and biohazard bags in the cleanroom or aseptic area. The nylon ties are long enough to easily fit around the neck of a 4 or 6 mil bag.

  • Product Type: Closure Ties
  • Material: Nylon
  • Color: White
  • Dimensions (L x W): 30.48 x 0.58 cm (11.75 x 0.23 in.)
  • Thickness: 0.13 cm (0.05 in.)
  • For Use With (Equipment): BinLiners™ and Biohazard Bags

Specifications

Autoclavable Autoclavable
Certifications/Compliance ISO 9001 Certified Facility
Closure Type Tie
Color White
Disposable Single-use
For Use With (Application) Bag Closure
For Use With (Equipment) BinLiners™ and Biohazard Bags
Material Nylon
No. per Case 25 x 4 Bags/Cs.
No. per Pack 5/Pk., 5 Pk./Double Bag
Sterilization Method Irradiated
Thickness (English) 0.05 in.
Dimensions (L x W) 30.48 x 0.58 cm (11.75 x 0.23 in.)
Thickness (Metric) 0.13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.