missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Closure Degassing

Product Code. 10591075
Click to view available options
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10591075

Brand: Kautex™ 2000085590

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Red
Includes Tamper Evident Ring and Venting Liner
Product Type Degassing Closure
For Use With (Equipment) 10 to 1000 mL Bottle
Diameter (Metric) 32 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.