missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Clostripain (Cl. Histolyticum)

Product Code. 16161760
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 16161760

Brand: Abnova™ P5214.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Name Clostripain (Cl. Histolyticum)
Preparation Method Native protein chromatographically purified from Clostridium histolyticum
Quantity 1 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name CLOSTRIPAIN
Assays Clostripain Assay; Peptide mapping; Sequence analysis; Cell isolation; Hydrolysis/condensation of amide bonds; Peptide synthesis.
Biological Activity ≥50 units per mg dry weight. One Unit hydrolyzes one micromole of N-benzoyl-L-arginine ethyl ester per minute at 25°C, pH 7.6, in the presence of dithiothreitol.
Species Clostridium species
Protein Tag None
Expression System Native protein chromatographically purified from Clostridium histolyticum
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.