missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Clostridium pasteurianum Rubredoxin Polyclonal Antibody
GREENER_CHOICE

Product Code. 17220124 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17220124 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17220124 Supplier Invitrogen™ Supplier No. PA5117623

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Rubredoxin is a small nonheme, iron protein lacking acid-labile sulfide. Its single Fe, chelated to 4 Cys, functions as an electron acceptor and may also stabilize the conformation of the molecule.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium pasteurianum Rubredoxin
Applications Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias Rubredoxin (Rd)
Host Species Rabbit
Immunogen Recombinant Clostridium pasteurianum Rubredoxin protein (1-54aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.