missing translation for 'onlineSavingsMsg'
Learn More

Clostridium Difficile Toxin A Antibody (actoxumab) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30161358
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30161358 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30161358 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328826JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

Clostridium Difficile Toxin A Monoclonal antibody specifically detects Clostridium Difficile Toxin A in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium Difficile Toxin A
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone actoxumab
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias tcdA, toxA
Host Species Human
Immunogen Cdiff Toxin A
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.