missing translation for 'onlineSavingsMsg'
Learn More

Cloprostenol, TRC

Product Code. 17972995
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972995 50 mg 50mg
17911815 10 mg 10mg
17921015 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17972995 Supplier Toronto Research Chemicals Supplier No. TRCC58730050MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40665-92-7
Molecular Formula C22 H29 Cl O6
Molecular Weight (g/mol) 424.92
Synonym 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-2-[(1E,3R)-4-(3-chlorophenoxy)-3-hydroxy-1-buten-1-yl]-3,5-dihydroxycyclopentyl]-, (5Z)-rel-, 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-2-[(1E,3R)-4-(3-chlorophenoxy)-3-hydroxy-1-butenyl]-3,5-dihydroxycyclopentyl]-, (5Z)-rel- (9CI), 5-Heptenoic acid, 7-[2-[4-(3-chlorophenoxy)-3-hydroxy-1-butenyl]-3,5-dihydroxycyclopentyl]-, [1α(Z),2β(1E,3R*),3α,5α]-(±)-, (±)-Cloprostenol, 5-Heptenoic acid, 7-[2-[4-(3-chlorophenoxy)-3-hydroxy-1-butenyl]-3,5-dihydroxycyclopentyl]-, [1α(Z),2β(1E,3R*),3α,5α]-, Cloprostenol, Clostenol, Cyclix, DL-Cloprostenol, Estrofan, Estromil, Estrophan, Estrophane, Estroplan, Juramate, Oestrophan, Oestrophane, Ovuprost, Racemic cloprostenol, rel-(5Z)-7-[(1R,2R,3R,5S)-2-[(1E,3R)-4-(3-Chlorophenoxy)-3-hydroxy-1-buten-1-yl]-3,5-dihydroxycyclopentyl]-5-heptenoic acid
IUPAC Name (Z)-7-[(1R,2R,3R,5S)-2-[(E,3R)-4-(3-chlorophenoxy)-3-hydroxybut-1-enyl]-3,5-dihydroxycyclopentyl]hept-5-enoic acid
SMILES O[C@@H](COc1cccc(Cl)c1)\C=C\[C@H]2[C@H](O)C[C@H](O)[C@@H]2C\C=C/CCCC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H29ClO6/c23-15-6-5-7-17(12-15)29-14-16(24)10-11-19-18(20(25)13-21(19)26)8-3-1-2-4-9-22(27)28/h1,3,5-7,10-12,16,18-21,24-26H,2,4,8-9,13-14H2,(H,27,28)/b3-1-,11-10+/t16-,18-,19-,20+,21-/m1/s1
Formula Weight 424.1653
Percent Purity >95%
Chemical Name or Material Cloprostenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.