missing translation for 'onlineSavingsMsg'
Learn More

Cloprostenol sodium salt, MedChemExpress

Product Code. 16462986
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462986 25 mg 25mg
16435156 100 mg 100mg
16424198 10 mg 10mg
16406917 5 mg 5mg
16400547 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16462986 Supplier Medchem Express Supplier No. HY108415
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cloprostenol sodium salt (ICI 80996 sodium salt) is a potent synthetic prostaglandin analogue, acts as a luteolytic agent, and is a PGF2α receptor agonist.

Cloprostenol sodium salt (ICI 80996 sodium salt) is a potent synthetic prostaglandin analogue, acts as a luteolytic agent, and is a PGF2α receptor agonist.

Specifications

Chemical Name or Material Cloprostenol sodium salt
CAS 55028-72-3
Color White
Purity Grade Notes Research
Molecular Formula C22H28ClNaO6
Quantity 25 mg
Synonym ICI 80996 sodium salt
Solubility Information H2O : ≥ 150 mg/mL (335.65 mM)
SMILES O=C(O[Na])CCC/C=C\C[C@H]1[C@H](/C=C/[C@H](O)COC2=CC=CC(Cl)=C2)[C@@H](O)C[C@H]1O
Molecular Weight (g/mol) 446.9
Formula Weight 446.9
Percent Purity 98.95%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.