missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clopidol, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Clopidol
Specifications
Specifications
| Chemical Name or Material | Clopidol |
| CAS | 2971-90-6 |
| Analyte or Component Names | Clopidol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H7 Cl2 N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2,6-Dimethyl-3,5-dichloro-4-pyridinol, 3,5-Dichloro-2,6-dimethyl-4-hydroxypyridine, 3,5-Dichloro-2,6-dimethyl-4-pyridinol, 3,5-Dichloro-2,6-dimethylpyridinol, 3,5-Dichloro-4-hydroxy-2,6-dimethylpyridine, Clopidol, Coccidiostat C, Coyden, Coyden 25, Farmcoccid, Lerbek, Methylchloropindol, Methylchlorpindol, Meticlorpindol, NSC 253479, Pharmcoccid, WR 61112 |
| InChI Formula | InChI=1S/C7H7Cl2NO/c1-3-5(8)7(11)6(9)4(2)10-3/h1-2H3,(H,10,11) |
| SMILES | Cc1nc(C)c(Cl)c(O)c1Cl |
| IUPAC Name | 3,5-dichloro-2,6-dimethylpyridin-4-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction