missing translation for 'onlineSavingsMsg'
Learn More

Clopidol, MedChemExpress

Product Code. 16408606
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16408606

Brand: Medchem Express HYB1088

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Clopidol (WR-61112) is an anticoccidial agent which is used as feed additive to control coccidiosis in chickens. Clopidol inhibits the sporulation of Eimeria tenella oocysts.

Clopidol (WR-61112) is an anticoccidial agent which is used as feed additive to control coccidiosis in chickens. Clopidol inhibits the sporulation of Eimeria tenella oocysts.

Specifications

Chemical Name or Material Clopidol
Physical Form Solid
CAS 2971-90-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H7Cl2NO
For Use With (Application) COVID-19-immunoregulation
Synonym WR-61112
Solubility Information 0.1 M NaOH : 26 mg/mL (135.39 mM; ultrasonic and adjust pH to 3 with NaOH) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES OC1=C(Cl)C(C)=NC(C)=C1Cl
Molecular Weight (g/mol) 192.04
Formula Weight 192.04
Percent Purity 99.9%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.