missing translation for 'onlineSavingsMsg'
Learn More

Clopidogrel thiolactone, MedChemExpress

Product Code. 16482947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482947 50 mg 50mg
16476888 10 mg 10mg
16459427 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16482947 Supplier Medchem Express Supplier No. HY15876
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Clopidogrel thiolactone is a P2Y12 receptor inhibitor, is a potent antiplatelet agent.

Clopidogrel thiolactone is a P2Y12 receptor inhibitor, is a potent antiplatelet agent.

Specifications

Chemical Name or Material Clopidogrel thiolactone
CAS 1147350-75-1
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C16H16ClNO3S
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (148.01 mM; Need ultrasonic)
SMILES O=C(OC)[C@@H](N1CCC(S2)C(C1)=CC2=O)C3=C(Cl)C=CC=C3
Molecular Weight (g/mol) 337.82
Formula Weight 337.82
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.