missing translation for 'onlineSavingsMsg'
Learn More

Clopidogrel, MedChemExpress

Product Code. 16486706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486706 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16486706 Supplier Medchem Express Supplier No. HY15283
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Clopidogrel is an orally active platelet inhibitor that targets P2Y12 receptor. Clopidogrel is used to inhibit blood clots in coronary artery disease, peripheral vascular disease, and cerebrovascular disease.

Clopidogrel is an orally active platelet inhibitor that targets P2Y12 receptor. Clopidogrel is used to inhibit blood clots in coronary artery disease, peripheral vascular disease, and cerebrovascular disease.

Specifications

Chemical Name or Material Clopidogrel
CAS 113665-84-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H16ClNO2S
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (155.37 mM; Need ultrasonic)
SMILES ClC1=C(C=CC=C1)[C@H](N2CCC3=C(C2)C=CS3)C(OC)=O
Molecular Weight (g/mol) 321.82
Formula Weight 321.82
Percent Purity 98.7%
Grade Research
Physical Form Viscous Liquid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.