missing translation for 'onlineSavingsMsg'
Learn More

Clopidogrel Hydrogen Sulphate, Mikromol™

Product Code. 17942836
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942836 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17942836 Supplier Mikromol Supplier No. MM0455.100250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Clopidogrel Hydrogen Sulphate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clopidogrel Hydrogen Sulfate
CAS 120202-66-6
Analyte or Component Names Clopidogrel Hydrogen Sulfate
Shipping Condition Room Temperature
Molecular Formula C16 H16 Cl N O2 S . H2 O4 S
Certifications/Compliance ISO 17034
Synonym Clopidogrel Hydrogen Sulfate, (S)-(+)-Methyl (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyrid-5-yl)acetate bisulfate, (S)-(+)-Methyl 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogen sulfate, Clopidogrel bisulfate, Clopidogrel hemisulfate, Clopidogrel hydrogen sulfate, Iscover, Methyl (S)-(4,5,6,7-tetrahydrothieno[3,2-c]pyridin-5-yl)(2-chlorophenyl)ethanoate hydrogen sulfate, Plavix, SR 25990C, (alphaS)-alpha-(2-Chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-acetic acid methyl ester sulfate (1:1), (S)-alpha-(2-Chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-acetic acid methyl ester sulfate (1:1)
InChI Formula InChI=1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)/t15-;/m0./s1
SMILES COC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl.OS(=O)(=O)O
IUPAC Name methyl (2S)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate;sulfuric acid
Molecular Weight (g/mol) 419.9
Quantity 250 mg
Formula Weight 419.0264 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.