missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clopidogrel Hydrogen Sulphate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Clopidogrel Hydrogen Sulfate |
| CAS | 120202-66-6 |
| Analyte or Component Names | Clopidogrel Hydrogen Sulfate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H16 Cl N O2 S . H2 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Clopidogrel Hydrogen Sulfate, (S)-(+)-Methyl (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyrid-5-yl)acetate bisulfate, (S)-(+)-Methyl 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogen sulfate, Clopidogrel bisulfate, Clopidogrel hemisulfate, Clopidogrel hydrogen sulfate, Iscover, Methyl (S)-(4,5,6,7-tetrahydrothieno[3,2-c]pyridin-5-yl)(2-chlorophenyl)ethanoate hydrogen sulfate, Plavix, SR 25990C, (alphaS)-alpha-(2-Chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-acetic acid methyl ester sulfate (1:1), (S)-alpha-(2-Chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-acetic acid methyl ester sulfate (1:1) |
| InChI Formula | InChI=1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)/t15-;/m0./s1 |
| Solution Type | Neat |
| SMILES | COC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl.OS(=O)(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?