missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (+)-Clopidogrel hydrogen sulfate

Description
Selective, high affinity P2Y12 receptor antagonist. Inhibits ADP-induced platelet aggregation and displays antithrombotic activity. Active enantiomer of (±)-clopidogrel hydrochloride (Cat. No. 2490).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 419.9 |
| Purity | >99% |
| Molecular Weight (g/mol) | 419.89 |
| Chemical Name or Material | (+)-Clopidogrel hydrogen sulfate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?