missing translation for 'onlineSavingsMsg'
Learn More

Clopidogrel hydrogen sulfate, MedChemExpress

Product Code. 16487316
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487316 100 mg 100mg
16411866 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16487316 Supplier Medchem Express Supplier No. HY17459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Clopidogrel hydrogen sulfate is an antiplatelet agent to prevent blood clots. Clopidogrel hydrogen sulfate inhibits CYP2B6 and CYP2C19 with IC50s of 18.2 nM and 524 nM, respectively. Clopidogrel hydrogen sulfate is a potent antithrombotic agent that inhibits ADP-induced platelet aggregation.Clopidogrel hydrogen sulfate also is an orally active P2Y(12) inhibitor.

Clopidogrel hydrogen sulfate is an antiplatelet agent to prevent blood clots. Clopidogrel hydrogen sulfate inhibits CYP2B6 and CYP2C19 with IC50s of 18.2 nM and 524 nM, respectively. Clopidogrel hydrogen sulfate is a potent antithrombotic agent that inhibits ADP-induced platelet aggregation.Clopidogrel hydrogen sulfate also is an orally active P2Y(12) inhibitor.

Specifications

Chemical Name or Material Clopidogrel hydrogen sulfate
CAS 120202-66-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H18ClNO6S2
Quantity 100 mg
Synonym (S)-(+)-Clopidogrel bisulfate (S)-(+)-Clopidogrel hydrogen sulfate
Solubility Information DMSO : ≥ 46.7 mg/mL (111.22 mM) ∣H2O : 16.67 mg/mL (39.70 mM; Need ultrasonic)
SMILES ClC1=C([C@@H](C(OC)=O)N2CCC3=C(C=CS3)C2)C=CC=C1.O=S(O)(O)=O
Molecular Weight (g/mol) 419.9
Formula Weight 419.9
Percent Purity 99.08%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.