missing translation for 'onlineSavingsMsg'
Learn More

Clonidine hydrochloride, MedChemExpress

Product Code. 16468286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468286 100 mg 100mg
16412126 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468286 Supplier Medchem Express Supplier No. HYB0409A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Clonidine hydrochloride is an agonist of α2-adrenoceptor and potent antihypertensive agent.

Clonidine hydrochloride is an agonist of α2-adrenoceptor and potent antihypertensive agent.

Specifications

Chemical Name or Material Clonidine hydrochloride
Physical Form Solid
CAS 4205-91-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H10Cl3N3
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 33.33 mg/mL (125.04 mM; Need ultrasonic) ∣DMSO : 7.6 mg/mL (28.51 mM; Need ultrasonic and warming)
SMILES ClC1=C(NC2=NCCN2)C(Cl)=CC=C1.Cl
Molecular Weight (g/mol) 266.55
Formula Weight 266.55
Percent Purity 99.89%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.