missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Clonetics™ MEBM™ Mammary Epithelial Cell Growth Basal Medium
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
missing translation for 'unitSize'
500mL
Description
The full MEGM™ BulletKit™ consists of basal medium and supplements. The MEGM™ BulletKit™ has been optimized for the growth of Mammary Epithelial cells in a serum-free environment. This media system has been referenced in over 130 unique publications primarily focusing on breast cancer research.
- The Basal Medium and SingleQuots™ Supplements and Growth Factors Kit can be purchased to replace and replenish components of the kit.
Specifications
Specifications
| Form | Liquid |
| Product Type | Aqueous solution |
| For Use With (Application) | Growth of mammary epithelial cells in a serum-free environment |
| Quantity | 500 mL |
| Format | Screw top bottle |
| pH | 7.25 to 7.65 |
| Content And Storage | Store in dark, 2° - 8°C |
| Description | Red |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction