Learn More
STEMCELL Technologies ClonaCell™-TCS/Hygromycin Kit
Complete kit containing ClonaCell™-TCS medium and hygromycin B antibiotic
Brand:  STEMCELL Technologies 03823
Additional Details : Weight : 0.31560kg
Product Code. 17138211
  • £756.00 / 1 set
Estimated Shipment: 01-05-2024
to see stock.
Description
Description
  • This kit is meant for selection and cloning applications where the hygromycin B antibiotic is used as the selective agent
  • ClonaCell™-TCS Medium is a serum-containing, methylcellulose-based semi-solid medium that can be used for selection and cloning of a variety of suspension-adapted and adherent cell lines, such as BHK-1, CHO-K1, CHO-S, CHO-DG44, and HEK-293
  • This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA)
  • Hygromycin B is suitable for use in cell culture for the selection and maintenance of hygromycin-resistant clones when using ClonaCell™-TCS Medium
Specifications
Specifications
CHO Cells, HEK-293,Other
Cell Culture; Semi-Solid Cloning
1 Kit