missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies ClonaCell™HY AOF Expansion Medium
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- ClonaCell™-HY AOF Expansion Medium is an animal origin-free (AOF) and serum-free liquid medium optimized for hybridoma expansion after hypoxanthine, aminopterin, thymidine (HAT) selection
- The medium contains hypoxanthine and thymidine (HT) and is used to wean hybridomas off aminopterin used during the selection process
- ClonaCell™-HY AOF Expansion Medium promotes robust expansion of hybridomas, adaptation from serum-containing to serum-free media, and stabilizing the viability and antibody productivity of hybridoma cell lines
- The majority of hybridomas can be switched directly from serum-containing media to ClonaCell™-HY AOF Expansion Medium without an adaptation step
- The absence of serum in this medium facilitates detection and purification of the desired hybridoma-derived antibody without interference from serum-derived immunoglobulins
Specifications
Specifications
| Cell Type | Hybridomas |
| For Use With (Application) | Cell Culture |
| Quantity | 500 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction