missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ClonaCell™ FLEX

Product Code. 17118211
Encompass
Click to view available options
Quantity:
45 mL
Unit Size:
45mL
This item is not returnable. View return policy

Product Code. 17118211

Brand: STEMCELL Technologies 03818

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Semi-solid methylcellulose-based matrix for selection and cloning of cells

  • ClonaCell™ FLEX is a base methylcellulose medium that requires the addition of a 2X concentrate of the user's own liquid medium to create a custom semi-solid cloning medium
  • This allows the user to adapt an existing liquid cloning method to a customized semi-solid cloning method
  • ClonaCell™ FLEX is recommended for selection and cloning of suspension-adapted mammalian cells, including CHO cells and hybridomas
  • The medium is chemically defined, animal component-free, and protein-free
  • It does not contain L-glutamine, selection agents, or phenol red
  • ClonaCell™ FLEX is compatible with a wide range of selection systems, including dihydrofolate reductase (DHFR), glutamine synthetase (GS), and hypoxanthine, aminopterin, thymidine (HAT)

Specifications

Cell Type Hybridomas; create dimCellLine - CHO Cells, HEK-293
For Use With (Application) Cell Culture; Semi-Solid Cloning
Quantity 45 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.