missing translation for 'onlineSavingsMsg'
Learn More

Clomipramine hydrochloride, Thermo Scientific™

Potent, selective 5-HT uptake blocker

Brand:  Thermo Scientific Alfa Aesar J62485.03


Quantity 1g

 View more versions of this product

Product Code. 15424869

  • £30.20 / 1g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

17321-77-6
351.315
WIMWMKZEIBHDTH-UHFFFAOYSA-N
68539
3-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
C19H24Cl2N2
MFCD00069234
clomipramine hydrochloride, anafranil, clomipramine hcl, 3-3-chloro-10,11-dihydro-5h-dibenzo b,f azepin-5-yl-n,n-dimethylpropan-1-amine hydrochloride, anaphranil, 3-chloroimipramine hydrochloride, chloroimipramine monohydrochloride, chlorimipramine hydrochloride, unii-2lxw0l6gwj
CHEBI:3755
CN(C)CCCN1C2=CC=CC=C2CCC3=C1C=C(C=C3)Cl.Cl

Specifications

17321-77-6
MFCD00069234
14,2387
WIMWMKZEIBHDTH-UHFFFAOYSA-N
3-(2-chloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
68539
351.32
C19H24Cl2N2
1g
clomipramine hydrochloride, anafranil, clomipramine hcl, 3-3-chloro-10,11-dihydro-5h-dibenzo b,f azepin-5-yl-n,n-dimethylpropan-1-amine hydrochloride, anaphranil, 3-chloroimipramine hydrochloride, chloroimipramine monohydrochloride, chlorimipramine hydrochloride, unii-2lxw0l6gwj
CN(C)CCCN1C2=CC=CC=C2CCC3=C1C=C(C=C3)Cl.Cl
351.315
CHEBI:3755
Clomipramine hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Clomipramine hydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H360-May damage fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Clomipramine hydrochloride

SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.