missing translation for 'onlineSavingsMsg'
Learn More

Clofibric acid, MedChemExpress

Product Code. 16438756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438756 Supplier Medchem Express Supplier No. HYB1415

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Clofibric acid (Chlorofibrinic acid), the pharmaceutically active metabolite of lipid regulators Clofibrate, Etofibrate and Etofyllinclofibrate, is a PPARα agonist which exhibits hypolipidemic effects. Clofibric acid also is an herbicide.

Clofibric acid (Chlorofibrinic acid), the pharmaceutically active metabolite of lipid regulators Clofibrate, Etofibrate and Etofyllinclofibrate, is a PPARα agonist which exhibits hypolipidemic effects. Clofibric acid also is an herbicide.

Specifications

Chemical Name or Material Clofibric acid
CAS 882-09-7
Color White
Purity Grade Notes Research
Molecular Formula C10H11ClO3
Quantity 100 mg
Synonym Chlorofibrinic acid
Solubility Information DMSO : ≥ 100 mg/mL (465.87 mM) ∣H2O : 1 mg/mL (4.66 mM; ultrasonic and warming and heat to 80°C)
SMILES CC(C)(OC1=CC=C(Cl)C=C1)C(O)=O
Molecular Weight (g/mol) 214.65
Formula Weight 214.65
Percent Purity 99.77%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.