missing translation for 'onlineSavingsMsg'
Learn More

Clofibric Acid Acyl-Beta-D-glucuronide (90%), TRC

Product Code. 30008169
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008169 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30008169 Supplier Toronto Research Chemicals Supplier No. TRCC5869401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72072-47-0
Molecular Formula C16 H19 Cl O9
Molecular Weight (g/mol) 390.77
Synonym Clofibric acid-acyl-beta-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-[2-(4-chlorophenoxy)-2-methylpropanoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CC(C)(Oc1ccc(Cl)cc1)C(=O)O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H19ClO9/c1-16(2,26-8-5-3-7(17)4-6-8)15(23)25-14-11(20)9(18)10(19)12(24-14)13(21)22/h3-6,9-12,14,18-20H,1-2H3,(H,21,22)/t9-,10-,11+,12-,14-/m0/s1
Formula Weight 390.0718
Chemical Name or Material Clofibric Acid Acyl-Beta-D-glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.