missing translation for 'onlineSavingsMsg'
Learn More

Clofibrate, Tocris Bioscience™

Product Code. 18742574
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18742574 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18742574 Supplier Tocris Bioscience Supplier No. 0824/1G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PPAR agonist

PPAR agonist (EC50 values are 50, 500 and > 100 μM at PPARα, PPARγ and PPARδ respectively). Antihyperlipoproteinemic.
TRUSTED_SUSTAINABILITY

Specifications

CAS 637-07-0
Molecular Formula C12H15ClO3
MDL Number MFCD00000615
Quantity 1 g
Synonym clofibrate, ethyl clofibrate, clofibratum, clofibrato, atromid-s, miscleron, atromid, liprin, epib, angiokapsul
InChI Key KNHUKKLJHYUCFP-UHFFFAOYSA-N
SMILES CCOC(=O)C(C)(C)OC1=CC=C(Cl)C=C1
IUPAC Name ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
Molecular Weight (g/mol) 242.70
PubChem CID 2796
ChEBI CHEBI:3750
Formula Weight 242.702
Percent Purity >98%
Chemical Name or Material Clofibrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.